Learn More
Anton Paar Kalliope 21 CFR Part 11 Module
Catalog No. 5000580488
Encompass_Preferred
Software module for Kalliope to comply with the regulation 21 CFRPart 11 introduced by US Food and Drug Administration (FDA)• Key features: electronic signature, audit trail, retrieval
Catalog No. 50-005-80488 Supplier Anton Paar Supplier No. 218627
Software module for Kalliope to comply with the regulation 21 CFRPart 11 introduced by US Food and Drug Administration (FDA)• Key features: electronic signature, audit trail, retrieval