Learn More
Anton Paar Kalliope Unlimited
Supplier:  Anton Paar 170678
Encompass_Preferred
Measurement series, databases for solvents, materials and SOPs are all easily accessible• Results from separate experiments can be compared and analyzed,and presented in customizable reports and/or exported to an Excel file for further processing and analysis• In combination with KALLIOPE PROFESSIONAL SOFTWAREMODULE 21 CFR PART 11 the software complies with the US FDA’sRegulation 21 CFR Part 11 concerning electronic records, audit trails and signatures• license for unlimited installations
Catalog No. 50-005-80528
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.