Learn More
Alkali Scientific Kanamycin Monosulfate, 10 G
SDP
Supplier:  Alkali Scientific KAN002CP
Encompass_Preferred
Kanamycin Monosulfate (10g) is an antibiotic used in cell culture selection to maintain cells that have incorporated the kanamycin resistance gene. This reagent is commonly used in genetic engineering studies, particularly in the selection and maintenance of transfected mammalian cells. Kanamycin is highly effective for selecting resistant cells, ensuring that only those expressing the desired gene will survive. This 10g powder form is ideal for preparing custom solutions for large-scale research projects, including gene expression studies, genetic modification, and pharmaceutical research. Kanamycin is a critical tool in molecular biology, biotechnology, and drug development.
Catalog No. 50-006-59717