Learn More
Alkali Scientific Kanamycin Monosulfate, 100 G
SDP
Supplier:  Alkali Scientific KAN003CP
Encompass_Preferred
Kanamycin Monosulfate is a broad-spectrum aminoglycoside antibiotic used for bacterial selection and research. It inhibits protein synthesis in susceptible bacteria by binding to the 30S ribosomal subunit, causing misreading of mRNA and disrupting bacterial growth. This formulation, containing 100g of Kanamycin Monosulfate, is ideal for use in cell culture systems and microbial research. It is commonly applied in selection procedures for resistant bacterial strains, especially in molecular biology, gene expression experiments, and antibiotic resistance studies. Kanamycin is particularly effective against Gram-negative bacteria and has been widely used in genetic research for decades.
Catalog No. 50-006-59897