Learn More
Gold Biotechnology Inc Kanamycin Monosulfate Solution
SDP
Supplier:  Gold Biotechnology Inc K120SL25
Encompass_Preferred
Kanamycin is an aminoglycoside antibiotic isolated from Streptomyces kanamyceticus It is effective against gram-positive and gram-negative bacteria as well as Mycoplasma species Kanamycin is commonly used to select for bacteria that have been transformed with a kanamycin-resistant gene and plant tissues that have the NPT II ( APH3 ) gene incorporated in them Kanamycin is very soluble in aqueous solution and is typically used in concentrations of 50-100 g/ml
Catalog No. 50-005-62058