Learn More
Kd Medical Inc KANAMYCIN SOLUTION 30MG/ML
SDP
Supplier:  Kd Medical Inc CAA1290
Encompass_Preferred
For selection for kanamycin resistance in mutated and transformed cells. One 10ml bottle.
Catalog No. 50-101-9045