Learn More
Alkali Scientific Kao & Michayluk Medium, Powder, 18.1g, 5 L
Supplier: Alkali Scientific KAO003CP
Kao & Michayluk Medium is a high-quality plant tissue culture medium available in powder form, making 5 liters of nutrient solution. This formulation supports the growth and development of a variety of plant species in laboratory conditions. It is frequently used in plant regeneration studies and for the culture of both herbaceous and woody plants. The medium provides essential vitamins, nutrients, and minerals that promote cell division and development in vitro. Kao & Michayluk medium is a crucial component in plant biotechnology for applications such as somatic embryogenesis, genetic studies, and tissue culture-based propagation of plants.