Learn More
Uline Kapton® Tape - 2 Mil, 1⁄4" x 36 yds
Supplier:  Uline S23760/DEL
Encompass
Maximum resistance to high temperatures.Recommended for masking circuit boards and splicing wires.Temperature range up to 500°F.Polyimide film with silicone adhesive for easy release.UL 510 flame retardant.
Catalog No. NC3522310