Learn More
Selleck Chemical LLC Kartogenin
SDP
Supplier:  Selleck Chemical LLC S765810MM/1ML
Encompass_Preferred
Kartogenin (KGN) is an activator of the smad4/smad5 pathway and promotes the selective differentiation of multipotent mesenchymal stem cells into chondrocytes
Catalog No. 50-000-00111