Learn More
TARGETMOL CHEMICALS INC Kasugamycin hydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T2762500MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Kasugamycin hydrochloride (Kasumin) is an aminoglycoside antibiotic that was originally isolated in 1965, from Streptomyces kasugaensis, a Streptomyces strain found near the Kasuga shrine in Nara, Japan. Kasugamycin hydrochloride was discovered by Hamao Umezawa, who also discovered kanamycin and bleomycin, as a drug which could prevent growth of a fungus causing rice blast disease. Purity 99.1%
Catalog No. 50-287-6525