Learn More
Medchemexpress LLC Katacalcin | 85916-47-8 | 98.3% | 2436.60 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP014910MG
Encompass_Preferred
Katacalcin is a potent plasma calcium-lowering peptide that belongs to the calcitonin family. It causes a rapid but short-lived drop in blood calcium and phosphate levels by promoting their incorporation into bones. It is also involved in regulating human CD14+ peripheral blood mononuclear cell (PBMC) migration.
  • Potent plasma calcium-lowering peptide.
  • Belongs to the calcitonin family.
  • Promotes the incorporation of calcium and phosphate into bones.
  • More potent inhibitor of fMLP-induced chemotaxis than calcitonin (CT) or procalcitonin (PCT) at specific concentrations.
  • Deactivates CD14+ PBMC chemotaxis towards various attractants.
  • Regulates human CD14+ PBMC migration via protein kinase A-dependent cAMP pathways.
  • High purity of 98.29%.
Catalog No. 50-004-25735