Learn More
Medchemexpress LLC Katacalcin (TFA) | 85916-47-8 | MFCD00133425 | 99.6% | 2550.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0149A5MG
Encompass_Preferred
Katacalcin TFA (PDN 21 TFA) is a potent peptide that lowers plasma calcium levels. It belongs to the calcitonin family, inducing a rapid, short-lived decrease in blood calcium and phosphate by promoting their bone incorporation.
  • Solid, white to off-white appearance
  • Potent plasma calcium-lowering peptide
  • Causes rapid, short-lived drop in blood calcium and phosphate levels
  • More potent inhibitor of fMLP-induced chemotaxis than calcitonin or procalcitonin
  • Deactivates CD14+ peripheral blood mononuclear cell (PBMC) chemotaxis
  • Regulates human CD14+ PBMC migration via protein kinase A-dependent cAMP pathways
Catalog No. 50-004-25894