Learn More
Cayman Chemical KatcalcIntrIfluoroacetate 10mg
SDP
Supplier:  Cayman Chemical 3928510 MG
Encompass_Preferred
An endogenous peptide; expressed in, and secreted from, thyroid parafollicular cells in humans; plasma levels are increased in patients with medullary thyroid carcinoma
Catalog No. 50-291-7214