Learn More
Medchemexpress LLC Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)- | 1220616-44-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN68331MG
Encompass_Preferred
Rebaudioside M is a non-calorie sweetener isolated from *Stevia rebaudiana*. It is a glycoside of the ent-kaurene diterpenoid aglycone and is more than 350 times sweeter than sucrose. It can be used in food and beverages.
  • Protects β-cell activity and enhances insulin secretion.
  • Inhibits PI3K/Akt pathway, lowering cell proliferation and enhancing apoptosis.
  • Downregulates Wnt/catenin pathway in colon cancer cells, reducing migration and invasion.
  • Lowers fasting blood glucose and enhances glucose tolerance in diabetic rats.
  • Decreases oxidative stress and pro-inflammatory cytokines, protecting pancreatic beta-cells.
  • Reduces blood pressure, increases endothelial function, and causes vasodilation in hypertensive rats.
Catalog No. 50-004-19521