Learn More
Medchemexpress LLC Kaur-16-ene-3,15-dione, 7,14-dihydroxy-, (7α,14R)- | 79498-31-0 | MFCD00915351 | 99.4% | 332.43 g/mol | C20H28O4 | 10 MG

Supplier: Medchemexpress LLC HYN211210MG
Glaucocalyxin A is an ent-kauranoid diterpene supplied for research use as an apoptosis inducer in biochemical and cell-based studies. It is provided as a white to off-white solid with high purity and is intended for laboratory research applications; follow recommended storage and solvent guidelines for handling and formulation.
- High purity (≈99.4%) suitable for research assays.
- Molecular weight 332.43 g/mol, formula C20H28O4.
- Supplied as a white to off-white solid for convenient weighing.
- Soluble in DMSO; sonication may be required for concentrated stocks.
- Powder stable at -20°C for extended storage.
- In vivo formulation protocols and solubilizing options provided for animal studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.