Learn More
Medchemexpress LLC Kavain | 3155-48-4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN209650MG
Encompass_Preferred
Kavain is a kavalactone isolated from *Piper methysticum*. It exhibits anxiolytic properties and acts as a positive modulator of γ-Aminobutyric acid type A (GABAA) receptors. This product is intended for research use.
  • Isolated from *Piper methysticum*.
  • Exhibits anxiolytic properties in animals and humans.
  • Positively modulates γ-Aminobutyric acid type A (GABAA) receptors.
  • Modulatory effect is unaffected by flumazenil.
  • Shows greater enhancement at α4β2δ than at α1β2γ2L GABAARs.
Catalog No. 50-004-21782