Learn More
TARGETMOL CHEMICALS INC Research Chemical 50mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T459050MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-Kavain ((R)-KAWAIN) is one of the six major kavalactones found in the Piper methysticum (kava) plant; reversible inhibitor of MAO-B. purity: 99%
Catalog No. 50-303-8147