Learn More
Selleck Chemical LLC Kavain S9409-1MG
SDP
Supplier:  Selleck Chemical LLC S94091MG
Encompass_Preferred
Kavain (( )-Kavain) the main kavalactone found mostly in the roots of the kava plant has anticonvulsive properties attenuating vascular smooth muscle contraction through interactions with voltage-dependent Na and Ca2 channels Kavain positively modulated -Aminobutyric acid type A (GABAA) receptor
Catalog No. 50-000-24532