Learn More
TARGETMOL CHEMICALS INC KB-0742 dihydrochloride 2MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T94462MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KB-0742 dihydrochloride is a potent, selective, and orally administered inhibitor of CDK9. Purity 99.32%
Catalog No. 50-287-9088