Learn More
Selleck Chemical LLC KB-0742 Dihydrochloride S9901-25MG
SDP
Supplier:  Selleck Chemical LLC S990125MG
Encompass_Preferred
KB-0742 Dihydrochloride is a potent selective and orally bioavailable small molecule inhibitor of the transcription elongation cofactor CDK9 with IC50 of 6 nM for CDK9/cyclin T1 inhibition at 10 M ATP
Catalog No. 50-000-24674