Learn More
SURE TECH DIAGNOSTIC ASSOC INC KB FETAL HEMOGLOBIN KT-ST101-A
Supplier:  SURE TECH DIAGNOSTIC ASSOC INC ST 101A
Encompass_Preferred
Includes 1 bottles each 101-10 Fixing Solution, 101-20 Citrate Buffer, 101-30 Staining Solution. Kleihauer Betke Staining Kit - For the identification of Fetal Eurythrocytes in Maternal Blood
Catalog No. NC9718282