Learn More
Cayman Chemical kb NB 142-70 25mg
SDP
Supplier:  Cayman Chemical 1800225 MG
Encompass_Preferred
A selective PKD inhibitor (IC50s = 28.3, 58.7, and 53.2 nM for PKD1, 2, and 3, respectively); inhibits prostate cancer cell migration and invasion, as well as reduces wound healing in vitro.
Catalog No. 50-298-0114