Learn More
Medchemexpress LLC KB SRC 4 | 1380088-03-8 | 99.2% | 555.03 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10848810MM/1ML
Encompass_Preferred
KB SRC 4 is a potent and highly selective c-Src inhibitor with antitumor activity. It shows no inhibition on c-Abl up to 125 μM.
  • Potent and highly selective c-Src inhibitor
  • Ki of 44 nM and Kd of 86 nM
  • Shows no inhibition on c-Abl up to 125 μM
  • Has antitumor activity
  • Selective over Lck, Fgr, c-Yes, Lyn, Hck, and Fyn family members
  • Inhibits growth of various cancer cell lines including HT-29, SK-BR-3, MCF7, MDA-MB-453, and NIH-3T3
Catalog No. 50-003-46356