Learn More
Medchemexpress LLC (-)-KCC2 blocker 1 | 1228439-73-3 | 98.24% | 415.50 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY18172B1MG
Encompass_Preferred
KCC2 blocker 1 is an orally active and selective K+-Cl- cotransporter KCC2 blocker with an IC50 of 1 μM. It is a benzyl prolinate. The product is for research use only and not sold to patients.
  • Appearance: Solid
  • Color: Light yellow to khaki
Catalog No. 50-003-87384