Learn More
Hach Company Filling Solution, Reference, 3 M KCl, 250 mL
Supplier:  Hach Company LZW9500.99
Encompass
Filling solution for electrode reference element, Potassium Chloride (KCl), 3 M, 250 mL bottle
Catalog No. 50-199-8014