Learn More
ABclonal Technology KCNJ12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143751000UL
Encompass_Preferred
This gene encodes an inwardly rectifying K+ channel which may be blocked by divalent cations. This protein is thought to be one of multiple inwardly rectifying channels which contribute to the cardiac inward rectifier current (IK1). The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Catalog No. 50-001-07714