Learn More
Aviva Systems Biology Corp KCNK4 Antibody - N-terminal region (ARP35380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KCNK4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KCNK4. Alternate names: K2p4.1, TRAAK, TRAAK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-55720