Learn More
Cayman Chemical KCNQ4 PotasIum Chnl Monc 100ug
SDP
Supplier:  Cayman Chemical 13713100 UG
Encompass_Preferred
Immunogen: Fusion protein amino acids 2-77 of human KCNQ • Host: Mouse • Species Reactivity: (+) Human, mouse, rat • Applications: ICC/IF, IHC, IP, WB
Catalog No. 50-295-3500