Learn More
Bioss KD-Validated AMBP Rabbit Recom
SDP
Supplier:  Bioss BSM90153R
Encompass_Preferred
Alpha-1-Microglobulin/Bikunin Precursor Primary Antibody Unconjugated Public Immunogen Range 50-100/352 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64917