Learn More
Bioss KD-Validated Atg16L1 Rabbit Re
SDP
Supplier:  Bioss BSM90168R
Encompass_Preferred
Autophagy Related 16 Like 1 Primary Antibody Unconjugated Public Immunogen Range 1-150/607 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-65916