Learn More
Bioss KD-Validated Beta-secretase 1
SDP
Supplier:  Bioss BSM90215R
Encompass_Preferred
Beta-Secretase 1 Primary Antibody Unconjugated Public Immunogen Range 450-501/501 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66485