Learn More
ABclonal Technology KD Validated BUB1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2319820UL
Encompass_Preferred
This gene encodes a serine threonine-protein kinase that play a central role in mitosis The encoded protein functions in part by phosphorylating members of the mitotic checkpoint complex and activating the spindle checkpoint This protein also plays a role in inhibiting the activation of the anaphase promoting complex cyclosome This protein may also function in the DNA damage response Mutations in this gene have been associated with aneuploidy and several forms of cancer Alternate splicing results in multiple transcript variants
Catalog No. 50-000-07869