Learn More
Bioss KD-Validated Catenin alpha 1 R
SDP
Supplier:  Bioss BSM90068R
Encompass_Preferred
Catenin Alpha 1 Primary Antibody Unconjugated Public Immunogen Range 1-65/906 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66847