Learn More
Bioss KD-Validated Desmin Rabbit Rec
SDP
Supplier:  Bioss BSM90407R
Encompass_Preferred
Desmin Primary Antibody Unconjugated Public Immunogen Range 80-130/375 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66299