Learn More
ABclonal Technology KD Validated ECHS1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A27295100UL
Encompass_Preferred
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs The gene product is a member of the hydratase isomerase superfamily It localizes to the mitochondrial matrix Transcript variants utilizing alternative transcription initiation sites have been described in the literature
Catalog No. 50-000-10326