Learn More
Bioss KD-Validated Eph receptor B1 R
SDP
Supplier:  Bioss BSM90233R
Encompass_Preferred
Ephrin type-B receptor 1 Primary Antibody Unconjugated Public Immunogen Range 575-625/984 Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66491