Learn More
Bioss KD-Validated Glyoxalase I Rabb
SDP
Supplier:  Bioss BSM90421R
Encompass_Preferred
Glyoxalase I Primary Antibody Unconjugated Public Immunogen Range 150-230/413 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67057