Learn More
ABclonal Technology KD Validated NF2/Merlin
SDP
Supplier:  ABclonal Technology A23186100UL
Encompass_Preferred
This gene encodes a protein that is similar to some members of the ERM (ezrin radixin moesin) family of proteins that link cytoskeletal components with proteins in the cell membrane The encoded protein is involved in regulation of contact-dependent inhibition of cell proliferation and functions in cell-cell adhesion and transmembrane signaling The encoded protein has been shown to interact with cell-surface proteins proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport Disruption of this protein s function has been implicated in tumorigenesis and metastasis Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities
Catalog No. 50-000-04937