Learn More
Bioss KD-Validated NR2C2 Rabbit Rabb
SDP
Supplier:  Bioss BSM90439R
Encompass_Preferred
Nuclear Receptor Subfamily 2 Group C Member 2 Primary Antibody Unconjugated Public Immunogen Range 250-313/313 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-67016