Learn More
Bioss KD-Validated Peptidase D Rabbi
SDP
Supplier:  Bioss BSM90126R
Encompass_Preferred
Peptidase D Primary Antibody Unconjugated Public Immunogen Range 461-493/493 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64909