Learn More
ABclonal Technology KD Validated PSAP Rabbit mAb
SDP
Supplier:  ABclonal Technology A22736100UL
Encompass_Preferred
This gene encodes a highly conserved preproprotein that is proteolytically processed to generate four main cleavage products including saposins A B C and D Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities Mutations in this gene have been associated with Gaucher disease and metachromatic leukodystrophy Alternative splicing results in multiple transcript variants at least one of which encodes an isoform that is proteolytically processed
Catalog No. 50-000-05703