Learn More
Bioss KD-Validated TAF15 Rabbit Reco
SDP
Supplier:  Bioss BSM90483R
Encompass_Preferred
TATA-Box Binding Protein Associated Factor 15 Primary Antibody Unconjugated Public Immunogen Range 15-77/510 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66931