Learn More
Bioss KD-Validated TARBP2 Rabbit Rec
SDP
Supplier:  Bioss BSM90509R
Encompass_Preferred
TARBP2 Subunit Of RISC Loading Complex Primary Antibody Unconjugated Public Immunogen Range 1-54/2799 Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-66979