Learn More
Medchemexpress LLC Kdm2b-in-4 | 1966933-85-6 | 99.6% | 376.49 | C24H28N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1396015MG
Encompass_Preferred
KDM2B-IN-4 is a potent small-molecule inhibitor of the histone demethylase KDM2B (compound 182b from patent WO2016112284A1). Supplied as a high-purity solid, the compound is intended for biochemical and cellular research into hyperproliferative diseases such as cancer and includes solubility and storage guidance for assay use.
  • Nanomolar potency against KDM2B (IC50 ≈ 1.12 nM).
  • High purity suitable for biochemical and cellular assays.
  • Available as a small-mass solid and as a DMSO solution for reconstitution.
  • High solubility in DMSO (up to 100 mg/mL) with in vivo formulation guidance.
  • Stable when stored as a powder at -20°C with multi-year shelf life.
Catalog No. 50-002-23228