Learn More
ABclonal Technology KDM7A Rabbit pAb
SDP
Supplier:  ABclonal Technology A146921000UL
Encompass_Preferred
Enables histone H3-methyl-lysine-9 demethylase activity and histone H3-tri/di-methyl-lysine-27 demethylase activity. Involved in histone H3-K27 demethylation, histone H3-K9 demethylation, and positive regulation of transcription, DNA-templated. Predicted to be located in nucleolus and nucleoplasm. Human ortholog(s) of this gene implicated in melanoma. Orthologous to human KDM7A (lysine demethylase 7A).
Catalog No. 50-001-07831