Learn More
Medchemexpress LLC KEA1-97 | 335.16 | C15H9Cl2FN4 | 100MG
SDP
Supplier:  Medchemexpress LLC HY114982100MG
Encompass_Preferred
KEA1-97 is a selective Thioredoxin-caspase 3 interaction disruptor (IC50 10 M) KEA1-97 disrupts the interaction of thioredoxin with caspase 3 activates caspases and induces apoptosis without affecting thioredoxin activity[1]
Catalog No. 50-002-28487