Learn More
Chemglass Life Sciences Clamp, Keck, Blue, Spherical Joint, Fits Joint Sizes 18/7 & 18/9
SDP
Supplier:  Chemglass Life Sciences CG145B21
Encompass_Preferred
Clamp, Keck, Blue, Spherical Taper, Fits Joint Sizes 18/7 & 18/9. Made of Delrin, these clamps are non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. CG145B21