Learn More
ATCC Keister's Modified TYI-S-33 Medium 1 vial prepares 15ml of medium
Supplier:  ATCC PRA2695
Encompass
Keister's Modified TYI-S-33 Giardia Medium [ATCC Medium 2695] is the recommended formulation for the axenic cultivation of Giardia and Spironucleus. It is provided freeze-dried, complete with 10% bovine serum, 1 vial prepares 15 mL of medium.
Catalog No. 50-238-3577