Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Kennedy Bin Holder Pegboard Hook, 4 in, Steel, 20 lb Capacity

Supplier:  MSC 00052472

Encompass

This bin holder pegboard hook is designed for organizing various bin sizes on a pegboard. Constructed from steel, it offers robust support for items up to 20 pounds, providing efficient storage for industrial and workshop environments.

  • Supports multiple bin sizes
  • Holds up to 20 lb
  • Made of steel

Catalog No. 50-005-29605


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.