Learn More
Selleck Chemical LLC Kenpaullone-5mg
SDP
Supplier:  Selleck Chemical LLC S79175MG
Encompass_Preferred
Kenpaullone (9-Bromopaullone, NSC-664704) is an ATP-competitive inhibitor of cyclin-dependent kinases (CDKs). It also inhibit glycogen synthase kinase 3β (GSK3β) with IC50 of 0.23 µM.
Catalog No. 50-238-9719